Skip to Content
MilliporeSigma

946168

Poly(ethylene glycol) methyl ether acrylate

Mn 2000, Tissue Engineering grade

Synonym(s):

Acrylic acid-methoxy(polyethylene glycol) ester, Methoxypolyethylene glycol acrylate, Methoxypolyethylene glycol monoacrylate, Methoxypolyoxyethylene acrylate, Oligo(ethylene oxide) methyl ether acrylate, PEG monoacrylate methyl ether, Poly(ethylene glycol) monomethyl ether monoacrylate, Poly(oligo(ethylene oxide) methyl ether acrylate), Polyethylene glycol acrylate methyl ether, Polyethylene glycol acrylate monomethyl ether, Polyethylene glycol methyl ether acrylate, Polyethylene glycol monomethyl ether acrylate, Polyethylene oxide methyl ether acrylate, Acryl-PEG, Methoxy PEG acrylate, Methoxy poly(ethylene glycol) monoacrylate, mPEG-acrylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

Linear Formula:
H2C=CHCO2(CH2CH2O)nCH3
CAS Number:
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

form

solid

mol wt

Mn 2000

color

white to light brown

Quality Level

particle size

>100 μm

density

1.09 g/mL at 25 °C (lit.), 1.09 g/mL

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Tissue Engineering (TE) Grade Poly(ethylene glycol) methyl ether acrylate (PEGMEA) is an inhibitor‑free, monofunctional acrylate macromer engineered for clean, rapid polymerization and biocompatible hydrogel environments. Developed for tissue engineering workflows,TE PEGMEA is R&D‑tested for low cytotoxicity leading to high cell viability. It features a ≥90% degree of functionalization (acrylate end‑group) to support reliable crosslinking and high conversion. The PEG backbone provides excellent aqueous solubility and biocompatibility, supporting scaffold fabrication, bioinks, encapsulation matrices, and protein‑resistant surface coatings.
Key Features
  • Inhibitor‑free formulation (no added MEHQ/BHT) for efficient curing and minimal residuals.
  • R&D‑tested biocompatibility demonstrating low cytotoxicity and high cell viability in standard lab assays. ≥90% degree of functionalization for consistent free‑radical polymerization and robust network formation.
  • PEG biocompatibility and hydrophilicity to help reduce protein adsorption and support gentle cell handling.
  • High purity and quality control suitable for demanding tissue engineering research and prototyping

Application

  1. Photocurable PEG hydrogels for scaffold fabrication and cell encapsulation
  2. Bioinks for 3D bioprinting (with suitable crosslinkers/initiators)
  3. Protein‑resistant surface coatings and soft network modifiers
  4. Drug‑delivery matrix components and permeability tuning

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service