Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

946192

Poly(ethylene glycol) dimethacrylate

Mn 10000, Tissue Engineering grade

Synonym(s):

Oligoethylene glycol dimethacrylate, Poly(ethylene oxide) dimethacrylate, Polyethylene glycol dimethacrylate, Polyethylene glycol methacrylate, Polyoxyethylene dimethacrylate, PEG dimethacrylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

Linear Formula:
C3H5C(O)(OCH2CH2)nOC(O)C3H5
CAS Number:
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI key

STVZJERGLQHEKB-UHFFFAOYSA-N

InChI

1S/C10H14O4/c1-7(2)9(11)13-5-6-14-10(12)8(3)4/h1,3,5-6H2,2,4H3

SMILES string

OCCO.CC(=C)C(O)=O

form

solid

mol wt

Mn 10000

color

white to light brown

bp

>200 °C/2 mmHg (lit.)

storage temp.

−20°C

Quality Level

Looking for similar products? Visit Product Comparison Guide

General description

Tissue Engineering Grade Poly(ethylene glycol) dimethacrylate (PEGDMA) is an inhibitor‑free, difunctional methacrylate macromer engineered for clean, rapid network formation and biocompatible hydrogel environments. Developed for tissue engineering workflows, TE PEGDMA is R&D‑tested for low cytotoxicity, supporting high cell viability under recommended formulation and curing conditions. It features a ≥90% degree of functionalization (dimethacrylate end‑groups) to enable reliable crosslinking and high conversion. The PEG backbone provides excellent aqueous solubility and biocompatibility, supporting scaffold fabrication, bioinks, encapsulation matrices, and protein‑resistant surface coatings.
Key Features:
  • Inhibitor‑free formulation (no added MEHQ/BHT) for efficient curing and minimal residuals.
  • R&D‑tested biocompatibility demonstrating low cytotoxicity and high cell viability in standard lab assays.
  • ≥90% degree of functionalization for consistent free‑radical polymerization and robust network formation.
  • PEG biocompatibility and hydrophilicity to help reduce protein adsorption and support gentle cell handling.
  • High purity and quality control suitable for demanding tissue engineering research and prototyping.


Application

  • Photocurable PEG hydrogels for scaffold fabrication and cell encapsulation
  • Bioinks for 3D bioprinting (with suitable crosslinkers/initiators)
  • Protein‑resistant surface coatings and soft network modifiers
  • Drug‑delivery matrix components and permeability tuning

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service