A60806

Sigma-Aldrich

2-Amino-6-methoxybenzothiazole

technical grade

Empirical Formula (Hill Notation):
C8H8N2OS
CAS Number:
Molecular Weight:
180.23
EC Number:
MDL number:
PubChem Substance ID:

grade

technical grade

mp

165-167 °C (lit.)

SMILES string

COc1ccc2nc(N)sc2c1

InChI

1S/C8H8N2OS/c1-11-5-2-3-6-7(4-5)12-8(9)10-6/h2-4H,1H3,(H2,9,10)

InChI key

KZHGPDSVHSDCMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

V Arjunan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 102, 327-340 (2012-12-12)
Extensive vibrational investigations of 2-amino-4-methoxybenzothiazole have been carried out with FTIR and FT-Raman spectral techniques. The electronic structure of the molecule has been analysed by UV-Visible and NMR spectroscopies. The DFT studies were carried out with B3LYP and HF methods...
Effect of Aroclor 1254 dose upon S9 activation of aromatic amine bacterial mutagens.
C Z Thompson et al.
Mutation research, 77(4), 361-366 (1980-04-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service