C69004

Sigma-Aldrich

3-Chloropropionaldehyde diethylacetal

95%

Synonym(s):
3-Chloro-1,1-diethoxypropane
Linear Formula:
ClCH2CH2CH(OC2H5)2
CAS Number:
Molecular Weight:
166.65
Beilstein/REAXYS Number:
1698609
EC Number:
MDL number:

assay

95%

contains

0.5% calcium carbonate as stabilizer

refractive index

n20/D 1.42 (lit.)

bp

84 °C/25 mmHg (lit.)

density

0.995 g/mL at 25 °C (lit.)

SMILES string

CCOC(CCCl)OCC

InChI

1S/C7H15ClO2/c1-3-9-7(5-6-8)10-4-2/h7H,3-6H2,1-2H3

InChI key

NXHONHDWVLPPCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN 1993C 3 / PGIII

WGK Germany

3

Flash Point(F)

129.2 °F

Flash Point(C)

54 °C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.