H6002

Sigma-Aldrich

Hexachlorocyclopentadiene

98%

Empirical Formula (Hill Notation):
C5Cl6
CAS Number:
Molecular Weight:
272.77
EC Number:
MDL number:
PubChem Substance ID:

vapor pressure

0.13 psi ( 20 °C)

assay

98%

refractive index

n20/D 1.5644 (lit.)

bp

239 °C/753 mmHg (lit.)

mp

−10 °C (lit.)

density

1.702 g/mL at 25 °C (lit.)

SMILES string

ClC1=C(Cl)C(Cl)(Cl)C(Cl)=C1Cl

InChI

1S/C5Cl6/c6-1-2(7)4(9)5(10,11)3(1)8

InChI key

VUNCWTMEJYMOOR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Jingwen Song et al.
The Science of the total environment, 748, 142525-142525 (2020-10-30)
Exposure to a single chemical does not exist in reality. Mixtures, which are the ecological norm, are often characterized by numerous intrinsic driving factors with unknown combined effects. Interactions between heterogeneous chemicals, or chemical and nonchemical stressors, could alter their...
Retention and fate of inhaled hexachlorocyclopentadiene in the rat.
L J Lawrence et al.
Bulletin of environmental contamination and toxicology, 26(5), 663-668 (1981-05-01)
J G Garcia et al.
Acta crystallographica. Section C, Crystal structure communications, 54 ( Pt 5), 645-647 (1998-06-24)
There are two independent molecules of endo-endo-anti-1,7,8,9,10,16,17,18,19,19,20,20-dodecachloro-4,13- dioxapentacyclo[14.2.1.1(7,10).0(2,15).0(6,11)]icosa-8,17-di ene, C18H12Cl12O2, in the unit cell with different conformations. In one, the ten-membered ring adopts a chair-chair conformation, and in the other, it adopts a distorted chair-chair conformation. There are near-zero torsion...
K M Abdo et al.
Journal of applied toxicology : JAT, 4(2), 75-81 (1984-04-01)
A 13-week subchronic study was conducted by administering hexachlorocyclopentadiene ( HCCP ) in corn oil by gavage to groups of ten male and ten female F344 rats at doses of 150, 75, 38, 19, 10 or 0 mg kg-1, and...
Fate of hexachlorocyclopentadiene in water and goldfish.
A A Podowski et al.
Archives of environmental contamination and toxicology, 13(4), 471-481 (1984-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service