MilliporeSigma
All Photos(1)

I1200

Sigma-Aldrich

Sodium iminodiacetate dibasic monohydrate

97%

Synonym(s):
Iminodiacetic acid disodium salt monohydrate
Linear Formula:
HN(CH2COONa)2 · H2O
CAS Number:
Molecular Weight:
195.08
Beilstein:
3790611
EC Number:
MDL number:

assay

97%

SMILES string

[Na+].[Na+].[H]O[H].[O-]C(=O)CNCC([O-])=O

InChI

1S/C4H7NO4.2Na.H2O/c6-3(7)1-5-2-4(8)9;;;/h5H,1-2H2,(H,6,7)(H,8,9);;;1H2/q;2*+1;/p-2

InChI key

YSBGCHXLMBAKPV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service