Documents

N17203

Sigma-Aldrich

3-Nitro-9-fluorenone

95%

Empirical Formula (Hill Notation):
C13H7NO3
CAS Number:
Molecular Weight:
225.20
MDL number:

assay

95%

SMILES string

[O-][N+](=O)c1ccc2C(=O)c3ccccc3-c2c1

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service