N17505

Sigma-Aldrich

5-Nitroindazole

97%

Empirical Formula (Hill Notation):
C7H5N3O2
CAS Number:
Molecular Weight:
163.13
Beilstein/REAXYS Number:
7936
EC Number:
MDL number:
PubChem Substance ID:

assay

97%

mp

204-208 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc2[nH]ncc2c1

InChI

1S/C7H5N3O2/c11-10(12)6-1-2-7-5(3-6)4-8-9-7/h1-4H,(H,8,9)

InChI key

WSGURAYTCUVDQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service