P3600

Sigma-Aldrich

Pentadecanoic acid

≥99%

Synonym(s):
Pentadecylic acid, NSC 28486, C15:0
Linear Formula:
CH3(CH2)13COOH
CAS Number:
Molecular Weight:
242.40
Beilstein/REAXYS Number:
1773831
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

assay

≥99%

form

powder, flakes, or chunks

bp

257 °C/100 mmHg (lit.)

mp

51-53 °C (lit.)

functional group

carboxylic acid

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCC(O)=O

InChI

1S/C15H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15(16)17/h2-14H2,1H3,(H,16,17)

InChI key

WQEPLUUGTLDZJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Flash Point(F)

>235.4 °F

Flash Point(C)

>113 °C

Mamoru Nanasato et al.
European journal of nuclear medicine and molecular imaging, 31(11), 1512-1521 (2004-07-03)
It is unclear whether 123I-labelled beta-methyl iodophenyl pentadecanoic acid (123I-BMIPP) myocardial scintigraphy adds further predictive value for future cardiac events compared with the variables obtained during cardiac catheterisation in patients with acute myocardial infarction (AMI) treated with primary percutaneous coronary...
Hidenori Seki et al.
Circulation journal : official journal of the Japanese Circulation Society, 69(3), 311-319 (2005-02-26)
(18)F-fluorodeoxyglucose (FDG) positron-emission tomography (PET) is assumed to be the most useful method of evaluating the viability of the myocardium, but its use is limited by the need for a cyclotron. In the present study, the ability of a combination...
M C Jong et al.
Diabetes, 50(12), 2779-2785 (2001-11-28)
Apolipoprotein (APO) C1 is a 6.6-kDa protein present in plasma and associated with lipoproteins. Using hyperinsulinemic-euglycemic clamp tests, we previously found that in APOC1 transgenic mice, the whole-body insulin-mediated glucose uptake is increased concomitant with a decreased fatty acid uptake....
A Wolk et al.
The Journal of nutrition, 131(3), 828-833 (2001-03-10)
The fatty acid intake is in part reflected by the fatty acid composition of adipose tissue and serum lipids. We evaluated whether the proportions of myristic (14:0), pentadecanoic (15:0) and heptadecanoic (17:0) fatty acids in the adipose tissue triacylglycerols and...
A Wolk et al.
The American journal of clinical nutrition, 68(2), 291-295 (1998-08-13)
We evaluated whether the adipose tissue content of 2 fatty acids of exogenous origin specific for ruminant fat, 15:0 and 17:0, reflect average long-term dairy fat consumption in free-living subjects. In 81 healthy women aged 30-77 y, we compared the...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.