Q809

Sigma-Aldrich

Quinaldine

≥95%

Synonym(s):
2-Methylquinoline
Empirical Formula (Hill Notation):
C10H9N
CAS Number:
Molecular Weight:
143.19
Beilstein/REAXYS Number:
110309
EC Number:
MDL number:
PubChem Substance ID:

Quality Level

assay

≥95%

refractive index

n20/D 1.612 (lit.)

bp

105-107 °C/10 mmHg (lit.)
248 °C (lit.)

mp

−9-−3 °C (lit.)

density

1.058 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc2ccccc2n1

InChI

1S/C10H9N/c1-8-6-7-9-4-2-3-5-10(9)11-8/h2-7H,1H3

InChI key

SMUQFGGVLNAIOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

H Cope et al.
European journal of medicinal chemistry, 41(10), 1124-1143 (2006-06-20)
Transmissible spongiform encephalopathies (TSEs) are thought to arise from aggregation of a protease resistant protein denoted PrP(Sc), which is a misfolded isoform of the normal cellular prion protein PrP(C). Using virtual high-throughput screening we have selected structures analogous to acridine...
J X Zhang et al.
Chemical senses, 30(9), 727-737 (2005-10-14)
Quantitative stir bar sorptive extraction methods, both in the aqueous and headspace modes, followed by thermal desorption gas chromatography-mass spectrometry were used to investigate individual variations in the volatile components of male and female ferret (Mustela furo) urine. The urinary...
Daniela Pucci et al.
Chemical communications (Cambridge, England), (19)(19), 2254-2256 (2008-05-09)
The synthesis of the first gallium(III)-based liquid crystal has been achieved grafting around the metal centre two chelating 2-methylquinolin-8-olate anions and one monodentate 3,4,5-tris(hexadecyloxy)benzoyloxy ligand, allowing the resulting complex to be a soft luminescent material with the typical high quantum...
J L Allen et al.
Veterinary and human toxicology, 28 Suppl 1, 21-24 (1986-01-01)
Residue concentrations of drugs that are administered to fish by bath immersion are related primarily to passage of the drugs across the gills. The elimination of these chemicals by fish can be mediated by biotransformation, but the route of elimination...
Terahertz absorption spectroscopy of a liquid using a polarity probe: a case study of trehalose/water mixtures.
Mohsen Sajadi et al.
Angewandte Chemie (International ed. in English), 49(2), 454-457 (2009-12-08)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service