MilliporeSigma
All Photos(1)

T38083

Sigma-Aldrich

o-Tolylacetic acid

ReagentPlus®, 99%

Synonym(s):
2-Methylphenylacetic acid
Linear Formula:
CH3C6H4CH2CO2H
CAS Number:
Molecular Weight:
150.17
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

product line

ReagentPlus®

assay

99%

mp

88-90 °C (lit.)

SMILES string

Cc1ccccc1CC(O)=O

InChI

1S/C9H10O2/c1-7-4-2-3-5-8(7)6-9(10)11/h2-5H,6H2,1H3,(H,10,11)

InChI key

RZWGTXHSYZGXKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

5 g in glass bottle
25 g in poly bottle

Legal Information

ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service