W215104

Sigma-Aldrich

Benzyl salicylate

≥98%, FG

Synonym(s):
Benzyl 2-hydroxybenzoate
Linear Formula:
2-(HO)C6H4CO2CH2C6H5
CAS Number:
Molecular Weight:
228.24
FEMA Number:
2151
Beilstein/REAXYS Number:
2115365
EC Number:
Council of Europe no.:
436
MDL number:
PubChem Substance ID:
Flavis number:
9.752
NACRES:
NA.21

Quality Level

biological source

synthetic

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

assay

≥98%

refractive index

n20/D 1.581 (lit.)

bp

168-170 °C/5 mmHg (lit.)

density

1.176 g/mL at 25 °C (lit.)

Documentation

see Safety & Documentation for available documents

Featured Industry

Flavors and Fragrances

Organoleptic

balsamic; sweet

food allergen

no known allergens

SMILES string

Oc1ccccc1C(=O)OCc2ccccc2

InChI

1S/C14H12O3/c15-13-9-5-4-8-12(13)14(16)17-10-11-6-2-1-3-7-11/h1-9,15H,10H2

InChI key

ZCTQGTTXIYCGGC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

H Nikawa et al.
Journal of oral rehabilitation, 22(11), 817-824 (1995-11-01)
Variation in the components of soft lining materials, i.e. the size of polymer particles, the ethyl alcohol content of the liquids and the type of plasticizer, were investigated with respect to their effects on the growth and colonization of Candida...
K A Kohrman et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 21(6), 741-744 (1983-12-01)
The potential of benzyl salicylate, an important fragrance and flavour ingredient, to induce hypersensitivity or to elicit reactions to pre-existing hypersensitivity in the general population was evaluated by analysing patch-test data. Results obtained from fragrance and formulator companies for a...
N Negreira et al.
Analytical and bioanalytical chemistry, 398(2), 995-1004 (2010-07-24)
The performance of the dispersive liquid-liquid microextraction (DLLME) technique for the determination of eight UV filters and a structurally related personal care species, benzyl salicylate (BzS), in environmental water samples is evaluated. After extraction, analytes were determined by gas chromatography...
D W Jones et al.
Journal of dental research, 65(5), 634-642 (1986-05-01)
An evaluation has been made of the soft polymer gel systems of the type used as prosthodontic short-term denture-lining materials. The purpose of the study was to determine the effect upon gel strength and gelation time produced by variations in...
Erik Christian Munksgaard
European journal of oral sciences, 112(1), 101-104 (2004-02-12)
The loss of the plasticizers dibutylphthalate, butylphthalylbutyl glycolate, benzylbenzoate, methylsalicylate, and benzylsalicylate from four soft lining materials was measured. A 0.1% aqueous solution of triton X-100, reduced was used as immersion medium, since the solubility of plasticizer in this medium...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service