W223115

Sigma-Aldrich

Camphor white oil

natural, FG

Synonym(s):
Cinnamomum Camphora Sieb, Camphora oil
CAS Number:
FEMA Number:
2231
Council of Europe no.:
0130
MDL number:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

FG
natural

Quality Level

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 172.510

refractive index

n20/D 1.468 (lit.)

bp

171 °C (lit.)

Documentation

see Safety & Documentation for available documents

Featured Industry

Flavors and Fragrances

Organoleptic

camphoraceous

food allergen

no known allergens

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

4 kg in steel drum
1 kg in glass bottle

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Hazard Codes

Xi

Risk Statement

10-36/38

Safety Statement

26

RIDADR

UN1130 - class 3 - PG 3 - Camphor oil

WGK Germany

WGK 2

Flash Point(F)

110.0 °F - closed cup

Flash Point(C)

43.33 °C - closed cup

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.