W231207

Sigma-Aldrich

Citronellyl butyrate

FCC, ≥90%

Linear Formula:
CH3CH2CH2CO2CH2CH2CH(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
226.36
FEMA Number:
2312
EC Number:
Council of Europe no.:
275
MDL number:
PubChem Substance ID:
Flavis number:
9.049

biological source

synthetic

Quality Level

reg. compliance

FCC

assay

≥90%

refractive index

n20/D 1.445 (lit.)

bp

245 °C (lit.)

density

0.873 g/mL at 25 °C (lit.)

Featured Industry

Flavors and Fragrances

SMILES string

CCCC(=O)OCCC(C)CC\C=C(\C)C

InChI

1S/C14H26O2/c1-5-7-14(15)16-11-10-13(4)9-6-8-12(2)3/h8,13H,5-7,9-11H2,1-4H3

InChI key

XQPZQXTWYZAXAK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service