W286818

Sigma-Aldrich

Phenethyl salicylate

≥97%, FG

Synonym(s):
Benzyl carbinyl salicylate
Linear Formula:
2-(HO)C6H4CO2CH2CH2C6H5
CAS Number:
Molecular Weight:
242.27
FEMA Number:
2868
EC Number:
Council of Europe no.:
437
MDL number:
PubChem Substance ID:
Flavis number:
9.753
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

biological source

synthetic

grade

FG
Kosher

assay

≥97%

bp

370 °C (lit.)

mp

39-41 °C (lit.)

density

1.154 g/mL at 25 °C (lit.)

Documentation

see Safety & Documentation for available documents

Featured Industry

Flavors and Fragrances

Organoleptic

carnation; hyacinth; balsamic; rose

food allergen

no known allergens

SMILES string

Oc1ccccc1C(=O)OCCc2ccccc2

InChI

1S/C15H14O3/c16-14-9-5-4-8-13(14)15(17)18-11-10-12-6-2-1-3-7-12/h1-9,16H,10-11H2

InChI key

YNMSDIQQNIRGDP-UHFFFAOYSA-N

Related Categories

Packaging

5, 10 kg in fiber drum
1 kg in poly bottle

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Legal Information

For R&D Use and/or Food Manufacturing only

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Target Organs

Respiratory system

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Hazard Codes

Xi

Risk Statement

36/37/38

Safety Statement

26-36

RIDADR

NONH for all modes of transport

WGK Germany

WGK 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.