MilliporeSigma
All Photos(1)

W295302

Sigma-Aldrich

1-Phenyl-2-pentanol

≥99%

Synonym(s):
α-Propylphenethyl alcohol
Empirical Formula (Hill Notation):
C11H16O
CAS Number:
Molecular Weight:
164.24
FEMA Number:
2953
EC Number:
Council of Europe no.:
83c
MDL number:
Flavis number:
2.034

assay

≥99%

SMILES string

CCCC(O)Cc1ccccc1

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Mild green, sweet, earthy

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service