W340103

Sigma-Aldrich

2-Heptylfuran

≥98%

Empirical Formula (Hill Notation):
C11H18O
CAS Number:
Molecular Weight:
166.26
FEMA Number:
3401
EC Number:
Council of Europe no.:
10952
MDL number:
PubChem Substance ID:
Flavis number:
13.069

grade

Halal
Kosher

assay

≥98%

refractive index

n20/D 1.45 (lit.)

bp

209-210 °C (lit.)

density

0.863 g/mL at 25 °C (lit.)

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

SMILES string

CCCCCCCc1ccco1

WGK Germany

3

Flash Point(F)

167 °F

Flash Point(C)

75 °C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.