W364908

Sigma-Aldrich

4-Propylphenol

≥97%, FG

Synonym(s):
p-Propylphenol
Linear Formula:
CH3CH2CH2C6H4OH
CAS Number:
Molecular Weight:
136.19
FEMA Number:
3649
EC Number:
MDL number:
PubChem Substance ID:
Flavis number:
4.050
NACRES:
NA.21

Quality Level

biological source

synthetic

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA

assay

≥97%

refractive index

n20/D 1.523 (lit.)

bp

232 °C (lit.)

density

0.983 g/mL at 25 °C (lit.)

Documentation

see Safety & Documentation for available documents

Featured Industry

Flavors and Fragrances

Organoleptic

medicinal

food allergen

no known allergens

SMILES string

CCCc1ccc(O)cc1

InChI

1S/C9H12O/c1-2-3-8-4-6-9(10)7-5-8/h4-7,10H,2-3H2,1H3

InChI key

KLSLBUSXWBJMEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

25 g in poly bottle
100 g in glass bottle
1 kg in glass bottle

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

Exclamation mark

Signal Word

Warning

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN 3145 8 / PGIII

WGK Germany

WGK 3

C J Rompelberg et al.
Chemico-biological interactions, 99(1-3), 85-97 (1996-01-05)
The irreversible and reversible inhibition of glutathione S-transferases (GSTs) by eugenol was studied in rat, mouse and man. Using liver cytosol of human, rat and mouse, species differences were found in the rate of irreversible inhibition of GSTs by eugenol...
Yuhe Liao et al.
Science (New York, N.Y.), 367(6484), 1385-1390 (2020-02-15)
The profitability and sustainability of future biorefineries are dependent on efficient feedstock use. Therefore, it is essential to valorize lignin when using wood. We have developed an integrated biorefinery that converts 78 weight % (wt %) of birch into xylochemicals....
Isabella Karlsson et al.
Photochemistry and photobiology, 88(4), 904-912 (2012-03-21)
Octocrylene is an organic UV filter, commonly used in sunscreens and cosmetics, which can give rise to both contact and photocontact allergy. Our aim was to investigate octocrylene's interaction with amino acid analogs in the presence of UV radiation to...
Shosuke Ito et al.
Journal of dermatological science, 80(1), 18-24 (2015-08-01)
Tyrosinase is able to oxidize a great number of phenols and catechols to form ortho-quinones. Ortho-quinones are highly reactive compounds that exert cytotoxicity through binding with thiol enzymes and the production of reactive oxygen species. Certain phenolic (and catecholic) compounds...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.