W379715

Sigma-Aldrich

4-Acetoxy-2,5-dimethyl-3(2H)furanone

natural (US), FG

Synonym(s):
Strawberry acetate
Empirical Formula (Hill Notation):
C8H10O4
CAS Number:
Molecular Weight:
170.16
FEMA Number:
3797
MDL number:
PubChem Substance ID:
Flavis number:
13.099
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

biological source

corn
sugar cane

grade

FG
Halal
Kosher
natural (US)

Agency/Method

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012

refractive index

n20/D 1.477 (lit.)

bp

243 °C (lit.)

density

1.167 g/mL at 25 °C (lit.)

Featured Industry

Flavors and Fragrances

Organoleptic

caramel; brown; sweet

food allergen

no known allergens

fragrance allergen

no known allergens

SMILES string

CC1OC(C)=C(OC(C)=O)C1=O

InChI

1S/C8H10O4/c1-4-7(10)8(5(2)11-4)12-6(3)9/h4H,1-3H3

InChI key

VPKIUOQJQJVLRW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Natural occurence: Yellow passion fruit, strawberry and microwave-blanched peanuts

Application

This compound enhances the profile of brown flavors, including caramel, butterscotch, molasses, brown sugar, chocolate mocha, coffee and toffee. It will add to the sweet caramelic notes of browned butter, custard, dulce de leche, strawberry and other fruit jams, BBQ, fried eggs and roasted meat and nuts and soy sauce flavors.

Packaging

100, 500 g in poly bottle

Biochem/physiol Actions

Taste at 20-40ppm

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Codes

Xn

Risk Statement

22

Safety Statement

36

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Organoleptic Characteristics of Flavor Materials
Michalski, J.
Perfumer & Flavorist, 39(4), 52-52 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.