MilliporeSigma
All Photos(1)

W429400

Sigma-Aldrich

Ethyl dimethyl dioxolane acetate

mixture of isomers, ≥97%, FG

Empirical Formula (Hill Notation):
C9H16O4
CAS Number:
Molecular Weight:
188.22
FEMA Number:
4294
EC Number:
MDL number:
PubChem Substance ID:
Flavis number:
6.087

biological source

synthetic

grade

FG

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

assay

≥97%

refractive index

n20/D 1.428 (lit.)

density

1.042 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

apple

SMILES string

CCOC(=O)CC1(C)OCC(C)O1

InChI

1S/C9H16O4/c1-4-11-8(10)5-9(3)12-6-7(2)13-9/h7H,4-6H2,1-3H3

InChI key

GSIXJEIRJVOUFB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service