W522600

Sigma-Aldrich

Opoponax oil

natural

Synonym(s):
Commiphora Erythraea
CAS Number:
MDL number:
NACRES:
NA.21
Pricing and availability is not currently available.

biological source

Commiphora erythrea

grade

Halal
Kosher
natural

origin

France origin

refractive index

n20/D 1.54 (lit.)

bp

238 °C/755 mmHg (lit.)

density

0.9 g/mL at 25 °C (lit.)

Documentation

see Safety & Documentation for available documents

Organoleptic

balsam

food allergen

no known allergens

Related Categories

Packaging

100 g in glass bottle
1 kg in glass bottle

Preparation Note

Extraction method: steam distillation

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

WGK Germany

WGK 3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.