MilliporeSigma
All Photos(1)

W526207

Sigma-Aldrich

Ethyl dimethyl dioxolane acetate

mixture of isomers

Empirical Formula (Hill Notation):
C9H16O4
CAS Number:
Molecular Weight:
188.22
EC Number:
MDL number:
PubChem Substance ID:
Flavis number:
6.087

refractive index

n20/D 1.428 (lit.)

density

1.042 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CC1(C)OCC(C)O1

InChI

1S/C9H16O4/c1-4-11-8(10)5-9(3)12-6-7(2)13-9/h7H,4-6H2,1-3H3

InChI key

GSIXJEIRJVOUFB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service