Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A55403

Sigma-Aldrich

3-Aminofluoranthene

90%

Synonym(s):

3-Fluoranthenamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11N
CAS Number:
Molecular Weight:
217.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

90%

mp

115-117 °C (lit.)

SMILES string

Nc1ccc2-c3ccccc3-c4cccc1c24

InChI

1S/C16H11N/c17-15-9-8-13-11-5-2-1-4-10(11)12-6-3-7-14(15)16(12)13/h1-9H,17H2

InChI key

VHGJAFIHUSTRGB-UHFFFAOYSA-N

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D L Berry et al.
Carcinogenesis, 6(10), 1403-1407 (1985-10-01)
1-Nitropyrene (1-NO2Py), 3-nitrofluoranthene (3-NO2Ft), 3-aminofluoranthene (3-NH2Ft) and 8-nitrofluoranthene (8-NO2Ft) were tested for mutagenicity in cultured Chinese hamster V79 cells. Mutations at the hypoxanthine-guanine phosphoribosyl transferase (HGPRT) gene locus were quantified. 1-NO2Py had marginal direct-acting mutagenicity which was enhanced by a
M A Belisario et al.
Toxicology, 108(1-2), 101-108 (1996-04-15)
Nitroarenes are environmental contaminants produced during incomplete combustion processes. Nitroreduction, the most important pathway of nitroarene toxification, occurs mainly in the liver and intestine. In the present study, we show that human red cells may also possess the metabolic competence
Nana Asare et al.
Toxicology, 255(3), 140-150 (2008-12-02)
In this study, we show that the environmental pollutant, 3-nitrofluoranthene (3-NF) but not its amine form, 3-aminofluoranthene (3-AF), induces apoptosis as well as regulated necrosis with necroptotic features in Hepa1c1c7 cells. Upon exposure to 3-NF, both typical apoptotic and necrotic
Claudia Dietze et al.
Analytical and bioanalytical chemistry, 407(29), 8735-8743 (2015-09-24)
A fast and straightforward method to prototype microfluidic chip systems for dead-volume-free hyphenation to electrospray-ionisation mass spectrometry is presented. The developed approach based on liquid-phase lithography provides an inexpensive and reliable access to microfluidic chips for MS coupling which can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service