MilliporeSigma
All Photos(1)

BR36008

BRAND® volumetric flask PP translucent

size 10 mL, joint: ST/NS 10/19, polypropylene stopper

Synonym(s):
Plastics, Volumetric flasks
NACRES:
NB.43

material

polypropylene stopper
translucent polypropylene flask

Joint

joint: ST/NS10/19

feature

accuracy class B

packaging

pack of 1 ea

manufacturer/tradename

BRAND 36008

size

10 mL

Related Categories

Compare Similar Items

View Full Comparison

1 of 4

This Item
BR36108BR37482BR37483
material

polypropylene stopper, translucent polypropylene flask

material

polypropylene stopper, translucent PMP flask

material

amber , borosilicate glass 3.3 flask, polypropylene stopper

material

amber , borosilicate glass 3.3 flask, polypropylene stopper

joint

joint: ST/NS10/19

joint

joint: ST/NS10/19

joint

joint ST/NS neck10/19, joint: 10/19

joint

joint ST/NS neck10/19, joint: 10/19

feature

accuracy class B

feature

accuracy class B

feature

class A DE-M marking. To DIN EN ISO 1042. Calibrated to contain (TC, In). With USP batch certificate.

feature

class A DE-M marking. To DIN EN ISO 1042. Calibrated to contain (TC, In). With USP batch certificate.

packaging

pack of 1 ea

packaging

pack of 1 ea

packaging

pack of 2 ea

packaging

pack of 2 ea

manufacturer/tradename

BRAND 36008

manufacturer/tradename

BRAND 36108

manufacturer/tradename

BRAND 37482

manufacturer/tradename

BRAND 37483

size

10 mL

size

10 mL

size

-

size

-

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

(enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service