Skip to Content
MilliporeSigma

C73605

2-Chloro-1,4-dimethylbenzene

99%

Synonym(s):

2-Chloro-p-xylene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(CH3)2C6H3Cl
CAS Number:
Molecular Weight:
140.61
EC Number:
202-444-2
UNSPSC Code:
12352100
MDL number:
Beilstein/REAXYS Number:
2040349
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

99%

refractive index

n20/D 1.524 (lit.)

bp

186 °C (lit.)

mp

2 °C (lit.)

density

1.049 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(C)c(Cl)c1

InChI

1S/C8H9Cl/c1-6-3-4-7(2)8(9)5-6/h3-5H,1-2H3

InChI key

KZNRNQGTVRTDPN-UHFFFAOYSA-N



pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

152.6 °F - closed cup

flash_point_c

67 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library