MilliporeSigma
All Photos(1)

Documents

D119407

Sigma-Aldrich

7-(2,3-Dihydroxypropyl)theophylline

99%

Sign Into View Organizational & Contract Pricing

Synonym(s):
7-(2,3-Dihydroxypropyl)-3,7-dihydro-1,3-dimethyl-1H-purine-2,6-dione, Diprophylline, Dyphylline
Empirical Formula (Hill Notation):
C10H14N4O4
CAS Number:
Molecular Weight:
254.24
Beilstein/REAXYS Number:
284563
EC Number:
MDL number:

assay

99%

mp

161-162 °C (lit.)

SMILES string

CN1C(=O)N(C)c2ncn(CC(O)CO)c2C1=O

InChI

1S/C10H14N4O4/c1-12-8-7(9(17)13(2)10(12)18)14(5-11-8)3-6(16)4-15/h5-6,15-16H,3-4H2,1-2H3

InChI key

KSCFJBIXMNOVSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Manthena V S Varma et al.
Journal of medicinal chemistry, 53(3), 1098-1108 (2010-01-15)
Oral bioavailability (F) is a product of fraction absorbed (Fa), fraction escaping gut-wall elimination (Fg), and fraction escaping hepatic elimination (Fh). In this study, using a database comprised of Fa, Fg, Fh, and F values for 309 drugs in humans
Xiaofan Sui et al.
European journal of medicinal chemistry, 44(11), 4455-4460 (2009-07-10)
We developed an improved Lombardeo's method (J. Med. Chem., 2004) for prediction of VD(ss) in human. With ElogD substituted by logk(IAM), together with f(i (7.4)) (the fraction of compound ionized at pH 7.4) and logf(u) (logarithmic fraction of compound unbound
R Scott Obach et al.
Drug metabolism and disposition: the biological fate of chemicals, 36(7), 1385-1405 (2008-04-23)
We present herein a compilation and trend analysis of human i.v. pharmacokinetic data on 670 drugs representing, to our knowledge, the largest publicly available set of human clinical pharmacokinetic data. This data set provides the drug metabolism scientist with a
Naomi L Kruhlak et al.
Toxicology mechanisms and methods, 18(2-3), 217-227 (2008-01-01)
ABSTRACT Drug-induced phospholipidosis (PL) is a condition characterized by the accumulation of phospholipids and drug in lysosomes, and is found in a variety of tissue types. PL is frequently manifested in preclinical studies and may delay or prevent the development

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service