Skip to Content
MilliporeSigma

GF01040645

Polyphenyleneoxide

(PPO ), (ppo (modified), ppe (modified)), rod, diam. 20 mm, L 200 mm, pale gray

Synonym(s):

PH307920

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

NACRES:
NA.23
UNSPSC Code:
12352100

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Polyphenyleneoxide, (PPO ), (ppo (modified), ppe (modified)), rod, diam. 20 mm, L 200 mm, pale gray

form

rod

manufacturer/tradename

Goodfellow 010-406-45

L

200 mm

diam.

20 mm

color

pale gray

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF52567016GF09505784GF43185056
diam.

20 mm

diam.

20 mm

diam.

20 mm

diam.

20 mm

L

200 mm

L

100 mm

L

1000 mm

L

500 mm

manufacturer/tradename

Goodfellow 010-406-45

manufacturer/tradename

Goodfellow 525-670-16

manufacturer/tradename

Goodfellow 095-057-84

manufacturer/tradename

Goodfellow 431-850-56

form

rod

form

rod

form

rod

form

rod

color

pale gray

color

pale gray

color

pale gray

color

pale gray

Legal Information

Product of Goodfellow

General description

For updated SDS information please visit www.goodfellow.com

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service