Skip to Content
MilliporeSigma

GF11444611

Gold

insulated wire, 5m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyimide insulation, 99.99%

Synonym(s):

Gold, AU005831

Sign In to View Organizational & Contract Pricing.

Select a Size

1 EA

$1,850.00

$1,850.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

About This Item

Empirical Formula (Hill Notation):
Au
CAS Number:
Molecular Weight:
196.97
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141717
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI

1S/Au

SMILES string

[Au]

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

assay

99.99%

form

wire

manufacturer/tradename

Goodfellow 114-446-11

resistivity

2.05 μΩ-cm, 0°C

L × diam. × thickness

5 m × 0.05 mm × 0.007 mm

bp

2808 °C (lit.)

mp

1063 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF88214542GF41388767GF06766713
Gold insulated wire, 5m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyimide insulation, 99.99%

GF11444611

Gold

Gold insulated wire, 1m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyimide insulation, 99.99%

GF88214542

Gold

Gold insulated wire, 2m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyimide insulation, 99.99%

GF41388767

Gold

Gold insulated wire, 5m, conductor diameter 0.125mm, insulation thickness 0.014mm, polyester insulation, 99.99%

GF06766713

Gold

assay

99.99%

assay

≥99.99%

assay

99.99%

assay

99.99%

manufacturer/tradename

Goodfellow 114-446-11

manufacturer/tradename

Goodfellow 882-145-42

manufacturer/tradename

Goodfellow 413-887-67

manufacturer/tradename

Goodfellow 067-667-13

form

wire

form

wire

form

wire

form

wire

density

19.3 g/mL at 25 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

bp

2808 °C (lit.)

bp

2808 °C (lit.)

bp

2808 °C (lit.)

bp

2808 °C (lit.)

mp

1063 °C (lit.)

mp

1063 °C (lit.)

mp

1063 °C (lit.)

mp

1063 °C (lit.)

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class

11 - Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Luo et al.
Journal of nanoscience and nanotechnology, 14(6), 4036-4042 (2014-04-18)
A novel immunosensor of human immune globulin (IgG) was fabricated based on the fluorescence transfer between luminescent semiconductor quantum dots (QDs) and gold nanoparticles (AuNPs). AuNPs and CdSe/ZnS QDs were respectively labeled with immune reaction pair:IgG and goat anti-human immunoglobulin
Jiajing Zhang et al.
Journal of nanoscience and nanotechnology, 14(6), 4124-4138 (2014-04-18)
Gold nanostructures with promising applications in biomedical field have attracted great attention. However, some fundamental questions other than the development of novel applications should be elucidated before they can actually serve as biomedicines in the clinic. Bio-safety is one of
Maria Jose Santos-Martinez et al.
Journal of biomedical nanotechnology, 10(6), 1004-1015 (2014-04-23)
The increasing use of gold nanoparticles in medical diagnosis and treatment has raised the concern over their blood compatibility. The interactions of nanoparticles with blood components may lead to platelet aggregation and endothelial dysfunction. Therefore, medical applications of gold nanoparticles
Cathleen M Crudden et al.
Nature chemistry, 6(5), 409-414 (2014-04-24)
Since the first report of thiol-based self-assembled monolayers (SAMs) 30 years ago, these structures have been examined in a huge variety of applications. The oxidative and thermal instabilities of these systems are widely known, however, and are an impediment to
Guojun Weng et al.
Journal of nanoscience and nanotechnology, 14(6), 4072-4077 (2014-04-18)
Gold nanorods of different aspect ratios had been synthesized using seed mediated growth method. The formed gold nanorods had been characterized by the absorption and transmission electron microscopy (TEM) measurements. The obtained gold nanorods were used to study the quenched

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service