MilliporeSigma
All Photos(2)

GF28613235

Iron

foil, 10mm disks, thickness 0.05mm, as rolled, 99.99+%

Sign Into View Organizational & Contract Pricing

Select a Size

Synonym(s):
Iron, FE000262
Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
MDL number:
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99%

form

foil

manufacturer/tradename

Goodfellow 286-132-35

resistivity

9.71 μΩ-cm

diam. × thickness

10 mm × 0.05 mm

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF15942618GF28380023GF92827413
Iron foil, 10mm disks, thickness 0.05mm, as rolled, 99.99+%

GF28613235

Iron

Iron foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.99+%

GF15942618

Iron

Iron foil, 4mm disks, thickness 0.05mm, as rolled, 99.99+%

GF28380023

Iron

Iron foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99.99+%

GF92827413

Iron

form

foil

form

foil

form

foil

form

foil

manufacturer/tradename

Goodfellow 286-132-35

manufacturer/tradename

Goodfellow 159-426-18

manufacturer/tradename

Goodfellow 283-800-23

manufacturer/tradename

Goodfellow 928-274-13

resistivity

9.71 μΩ-cm

resistivity

9.71 μΩ-cm

resistivity

9.71 μΩ-cm

resistivity

9.71 μΩ-cm

bp

2750 °C (lit.)

bp

2750 °C (lit.)

bp

2750 °C (lit.)

bp

2750 °C (lit.)

mp

1535 °C (lit.)

mp

1535 °C (lit.)

mp

1535 °C (lit.)

mp

1535 °C (lit.)

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Qiu et al.
Physical review letters, 85(7), 1492-1495 (2000-09-06)
FeF (2) films are grown by the reaction of XeF (2) and SeF (6) with iron foil. The growth initially follows the Mott-Cabrera parabolic rate law, indicating that the process is diffusion limited. At a certain film thickness, however, the
Jun-Won Jang et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 59(12), 2503-2507 (2009-06-23)
Zero valent iron has been successfully used for the degradation of a wide range of contaminants. However, this reaction of using ZVI particle produces a large quantity of iron sludge. To solve the problem, we report the synthesis of self-organized
Thomas A Russo et al.
Infection and immunity, 82(6), 2356-2367 (2014-03-26)
Hypervirulent (hypermucoviscous) Klebsiella pneumoniae (hvKP) strains are an emerging variant of "classical" K. pneumoniae (cKP) that cause organ and life-threatening infection in healthy individuals. An understanding of hvKP-specific virulence mechanisms that enabled evolution from cKP is limited. Observations by our
Hypoferremia of infection: a double-edged sword?
Kristen L Lokken et al.
Nature medicine, 20(4), 335-337 (2014-04-09)
Eunna Choi et al.
Infection and immunity, 82(6), 2203-2210 (2014-03-20)
In order to survive inside macrophages, Salmonella produces a series of proteins encoded by genes within Salmonella pathogenicity island 2 (SPI-2). In the present study, we report that Fur, a central regulator of iron utilization, negatively controls the expression of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service