Skip to Content
MilliporeSigma

GF31900508

Nickel

foil, 50x50mm, thickness 0.125mm, as rolled, 99.99+%

Synonym(s):

Nickel, NI000520

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141729
MDL number:
eCl@ss:
38190701

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI

1S/Ni

SMILES string

[Ni]

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

assay

99.99%

form

foil

manufacturer/tradename

Goodfellow 319-005-08

resistivity

6.97 μΩ-cm, 20°C

size × thickness

50x50 mm × 0.125 mm

bp

2732 °C (lit.)

mp

1453 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF32919824GF82377816GF72359017
manufacturer/tradename

Goodfellow 319-005-08

manufacturer/tradename

Goodfellow 329-198-24

manufacturer/tradename

Goodfellow 823-778-16

manufacturer/tradename

Goodfellow 723-590-17

form

foil

form

foil

form

foil

form

foil

assay

99.99%

assay

99.99%

assay

99.999%

assay

99.999%

resistivity

6.97 μΩ-cm, 20°C

resistivity

6.97 μΩ-cm, 20°C

resistivity

6.97 μΩ-cm, 20°C

resistivity

6.97 μΩ-cm, 20°C

mp

1453 °C (lit.)

mp

1453 °C (lit.)

mp

1453 °C (lit.)

mp

1453 °C (lit.)

bp

2732 °C (lit.)

bp

2732 °C (lit.)

bp

2732 °C (lit.)

bp

2732 °C (lit.)

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Carc. 2 - Skin Sens. 1 - STOT RE 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Kim et al.
Applied physics letters, 98(26), 263106-2631063 (2011-07-30)
Microwave plasma chemical vapor deposition (MPCVD) was employed to synthesize high quality centimeter scale graphene film at low temperatures. Monolayer graphene was obtained by varying the gas mixing ratio of hydrogen and methane to 80:1. Using advantages of MPCVD, the
H T Tien et al.
Photochemistry and photobiology, 49(5), 527-530 (1989-05-01)
Visible light of the solar spectrum is directly converted to stored chemical energy of hydrogen from artificial sea water in a novel electrochemical photovoltaic cell. The principal element of the cell, modeled after the photosynthetic thylakoid membrane, is a semiconductor
L A van Loon et al.
Journal of oral pathology, 17(3), 129-137 (1988-03-01)
An in vivo comparison was made between the contact allergic stomatitis-inducing capacity of nickel, nickel-containing dental alloys and a non-corrosive precious metal. Fifteen patients with a positive allergic skin reaction to nickel were divided into 3 groups (A, B and
Gareth E Haslam et al.
Physical chemistry chemical physics : PCCP, 13(28), 12968-12974 (2011-06-23)
Metallic nickel is a powerful electrocatalyst in alkaline solution and is able to be used in the alkaline fuel cell. However, in acidic solution, electrocatalysis is impossible because the metal is subject to rapid corrosion at low pH for all
G E van Dorssen et al.
Journal of X-ray science and technology, 3(2), 109-117 (1992-01-01)
The structure of Ni-C multilayer and single nickel layer samples has been analyzed before and after annealing, using two techniques: fluorescence EXAFS (F1EXAFS) at the Ni-K. edge and CuKα reflection. Annealing at a temperature of 450°C resulted in a change

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service