Skip to Content
MilliporeSigma

GF38604854

Brass alloy, Cu63Zn37

100mm tube, 1.5mm outside diameter, 0.25mm wall thickness, 1mm inside diameter, as drawn

Synonym(s):

CU027200

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

UNSPSC Code:
30102010
NACRES:
NA.23
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

form

tubes

manufacturer/tradename

Goodfellow 386-048-54

L × wall thickness

100 mm × 0.25 mm

O.D. × I.D.

1.5 mm × 1 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service