MilliporeSigma
All Photos(1)

GF43173581

Polyetherimide

(PEI), sheet, thickness 1.6 mm, size 600 × 600 mm, clear amber

Synonym(s):
EI313020
NACRES:
NA.23

form

sheet

manufacturer/tradename

Goodfellow 431-735-81

size

600 × 600 mm

thickness

1.6 mm

color

clear amber

Related Categories

Compare Similar Items

View Full Comparison

1 of 4

This Item
GF52105621GF22252430GF65554612
vibrant-m

GF43173581

Polyetherimide

vibrant-m

GF52105621

Polyetherimide

vibrant-m

GF22252430

Polyetherimide

vibrant-m

GF65554612

Polyetherimide

form

sheet

form

rod

form

rod

form

film

manufacturer/tradename

Goodfellow 431-735-81

manufacturer/tradename

Goodfellow 521-056-21

manufacturer/tradename

Goodfellow 222-524-30

manufacturer/tradename

Goodfellow 655-546-12

thickness

1.6 mm

thickness

-

thickness

-

thickness

0.5 mm

size

1.6 mm

size

-

size

-

size

0.5 mm

color

clear amber

color

clear amber

color

clear amber

color

clear amber

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service