GF44138268
99%
foil
Goodfellow 441-382-68
126 μΩ-cm, 20°C
10 × 10 mm
2.0 mm
3273 °C (lit.)
1313 °C (lit.)
7.886 g/mL at 25 °C (lit.)
[Gd]
1S/Gd
UIWYJDYFSGRHKR-UHFFFAOYSA-N
Danger
Flam. Sol. 1 - Water-react. 3
4.3 - Hazardous materials, which set free flammable gases upon contact with water
WGK 3
Not applicable
Not applicable
Enter Lot Number to search for Certificate of Analysis (COA).
Enter Lot Number to search for Certificate of Origin (COO).