MilliporeSigma
All Photos(2)

GF49978029

Europium

foil, 50x50mm, thickness 0.1mm, as rolled, 99.95%

Synonym(s):
Europium, EU000200
Empirical Formula (Hill Notation):
Eu
CAS Number:
Molecular Weight:
151.96
MDL number:
PubChem Substance ID:
NACRES:
NA.23

assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 499-780-29

resistivity

81 μΩ-cm, 20°C

size × thickness

50 x 50 mm × 0.1 mm

bp

1527 °C (lit.)

mp

822 °C (lit.)

density

5.24 g/mL at 25 °C (lit.)

SMILES string

[Eu]

InChI

1S/Eu

InChI key

OGPBJKLSAFTDLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com.

Packaging

1 ea

Legal Information

Product of Goodfellow

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service