Skip to Content
MilliporeSigma

GF54738760

Alumina

tube, outside diameter 4.0 mm, length 150 mm, wall thickness 1.0 mm

Synonym(s):

AL607080

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

NACRES:
NA.23
UNSPSC Code:
11181511

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

form

tubes

manufacturer/tradename

Goodfellow 547-387-60

L

150 mm

O.D.

4.0 mm

wall thickness

1.0 mm

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF06853855GF71365451GF51152475
form

tubes

form

tubes

form

tubes

form

tubes

L

150 mm

L

150 mm

L

150 mm

L

100 mm

wall thickness

1.0 mm

wall thickness

1.5 mm

wall thickness

0.6 mm

wall thickness

1.5 mm

manufacturer/tradename

Goodfellow 547-387-60

manufacturer/tradename

Goodfellow 068-538-55

manufacturer/tradename

Goodfellow 713-654-51

manufacturer/tradename

Goodfellow 511-524-75

O.D.

4.0 mm

O.D.

10.0 mm

O.D.

2.5 mm

O.D.

8.0 mm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service