Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF56814903

Brass alloy, Cu63Zn37

foil, 10m coil, 0.025mm thickness, hard

Synonym(s):

CU020220

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$939.00

$939.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
$939.00

About This Item

UNSPSC Code:
30102010
NACRES:
NA.23

$939.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

form

foil (coil - hard)

manufacturer/tradename

Goodfellow 568-149-03

size × thickness

10 m × 0.025 mm

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF00859169GF06322104GF90263617
form

foil (coil - hard)

form

foil (coil - hard)

form

foil (coil - hard)

form

foil (coil - hard)

manufacturer/tradename

Goodfellow 568-149-03

manufacturer/tradename

Goodfellow 008-591-69

manufacturer/tradename

Goodfellow 063-221-04

manufacturer/tradename

Goodfellow 902-636-17

size × thickness

10 m × 0.025 mm

size × thickness

1 m × 0.025 mm

size × thickness

1 m × 0.038 mm

size × thickness

2 m × 0.038 mm

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service