Skip to Content
MilliporeSigma

GF58250099

Zirconium

foil, 50mm disks, thickness 0.02mm, 99.2%

Synonym(s):

Zirconium, ZR000200

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
Zr
CAS Number:
Molecular Weight:
91.22
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141751
MDL number:
eCl@ss:
38180201
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

99.2%

form

foil

manufacturer/tradename

Goodfellow 582-500-99

resistivity

40 μΩ-cm, 20°C

size × thickness

50x50 mm × 0.175 mm

bp

4377 °C (lit.)

mp

1852 °C (lit.)

density

6.5 g/mL at 25 °C (lit.)

SMILES string

[Zr]

InChI

1S/Zr

InChI key

QCWXUUIWCKQGHC-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow


Still not finding the right product?

Explore all of our products under Zirconium


Storage Class

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library