Skip to Content
MilliporeSigma

GF73082976

Iron

rod, 100mm, diameter 80mm, as drawn, armcO« soft ingot 99.8%

Synonym(s):

Iron, FE007990

Slide 1 of 2
Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$731.00

$731.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

About This Item

Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
MDL number:
UNSPSC Code:
12141721
PubChem Substance ID:
NACRES:
NA.23

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

assay

99.8%

form

rod

manufacturer/tradename

Goodfellow 730-829-76

resistivity

9.71 μΩ-cm

L × diam.

100 mm × 80 mm

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF42842336GF35426291GF81029831
Iron rod, 100mm, diameter 80mm, as drawn, armcO« soft ingot 99.8%

GF73082976

Iron

Iron rod, 100mm, diameter 40mm, as drawn, armcO« soft ingot 99.8%

GF42842336

Iron

Iron rod, 50mm, diameter 80mm, as drawn, armcO« soft ingot 99.8%

GF35426291

Iron

Iron rod, 100mm, diameter 50mm, as drawn, armcO« soft ingot 99.8%

GF81029831

Iron

assay

99.8%

assay

99.8%

assay

99.8%

assay

99.8%

manufacturer/tradename

Goodfellow 730-829-76

manufacturer/tradename

Goodfellow 428-423-36

manufacturer/tradename

Goodfellow 354-262-91

manufacturer/tradename

Goodfellow 810-298-31

form

rod

form

rod

form

rod

form

rod

mp

1535 °C (lit.)

mp

1535 °C (lit.)

mp

1535 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

resistivity

9.71 μΩ-cm

resistivity

9.71 μΩ-cm

resistivity

9.71 μΩ-cm

resistivity

9.71 μΩ-cm

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

50 years ago in the Journal of Pediatrics: Iron metabolism in premature infants: II. Prevention of iron deficiency.
Pamela J Kling
The Journal of pediatrics, 164(4), 768-768 (2014-03-22)
Thomas A Russo et al.
Infection and immunity, 82(6), 2356-2367 (2014-03-26)
Hypervirulent (hypermucoviscous) Klebsiella pneumoniae (hvKP) strains are an emerging variant of "classical" K. pneumoniae (cKP) that cause organ and life-threatening infection in healthy individuals. An understanding of hvKP-specific virulence mechanisms that enabled evolution from cKP is limited. Observations by our
Laura M van Staalduinen et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(14), 5171-5176 (2014-04-08)
The enzymes PhnY and PhnZ comprise an oxidative catabolic pathway that enables marine bacteria to use 2-aminoethylphosphonic acid as a source of inorganic phosphate. PhnZ is notable for catalyzing the oxidative cleavage of a carbon-phosphorus bond using Fe(II) and dioxygen
Eunna Choi et al.
Infection and immunity, 82(6), 2203-2210 (2014-03-20)
In order to survive inside macrophages, Salmonella produces a series of proteins encoded by genes within Salmonella pathogenicity island 2 (SPI-2). In the present study, we report that Fur, a central regulator of iron utilization, negatively controls the expression of
Joseph D Mancias et al.
Nature, 509(7498), 105-109 (2014-04-04)
Autophagy, the process by which proteins and organelles are sequestered in double-membrane structures called autophagosomes and delivered to lysosomes for degradation, is critical in diseases such as cancer and neurodegeneration. Much of our understanding of this process has emerged from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service