Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF76657203

Iron

rod, 100mm, diameter 9.5mm, as drawn, soft ingot 99.8+%

Synonym(s):

Iron, FE007951

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$278.00

$278.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
$278.00

About This Item

Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
MDL number:
UNSPSC Code:
12141721
PubChem Substance ID:
NACRES:
NA.23

$278.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

assay

99.8%

form

rod

manufacturer/tradename

Goodfellow 766-572-03

resistivity

9.71 μΩ-cm

L × diam.

100 mm × 9.5 mm

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF89491045GF91830359GF02974226
Iron rod, 100mm, diameter 9.5mm, as drawn, soft ingot 99.8+%

GF76657203

Iron

Iron rod, 1000mm, diameter 9.5mm, as drawn, soft ingot 99.8+%

GF89491045

Iron

Iron rod, 100mm, diameter 19mm, as drawn, soft ingot 99.8%

GF91830359

Iron

Iron rod, 500mm, diameter 6.35mm, as drawn, soft ingot 99.8%

GF02974226

Iron

assay

99.8%

assay

≥99.8%

assay

≥99.8%

assay

99.80%

manufacturer/tradename

Goodfellow 766-572-03

manufacturer/tradename

Goodfellow 894-910-45

manufacturer/tradename

Goodfellow 918-303-59

manufacturer/tradename

Goodfellow 029-742-26

form

rod

form

rod

form

rod

form

rod

mp

1535 °C (lit.)

mp

1535 °C (lit.)

mp

1535 °C (lit.)

mp

1535 °C (lit.)

resistivity

9.71 μΩ-cm

resistivity

9.71 μΩ-cm

resistivity

9.71 μΩ-cm

resistivity

9.71 μΩ-cm

bp

2750 °C (lit.)

bp

2750 °C (lit.)

bp

2750 °C (lit.)

bp

2750 °C (lit.)

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service