MilliporeSigma
  • GF77250865
All Photos(2)

GF77250865

Zinc

foil, 4mm disks, thickness 0.025mm, as rolled, 99.95+%

Synonym(s):
Zinc, ZN000220
Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
PubChem Substance ID:

vapor pressure

1 mmHg ( 487 °C)

Assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 772-508-65

resistivity

5.8 μΩ-cm, 20°C

diam. × thickness

4 mm × 0.025 mm

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

1 of 4

This Item
GF70562112GF13982643GF37917232
Zinc foil, 4mm disks, thickness 0.025mm, as rolled, 99.95+%

GF77250865

Zinc

Zinc foil, 4mm disks, thickness 0.075mm, as rolled, 99.95+%

GF70562112

Zinc

Zinc foil, 4mm disks, thickness 0.125mm, as rolled, 99.95+%

GF13982643

Zinc

Zinc foil, 4mm disks, thickness 0.20mm, as rolled, 99.95+%

GF37917232

Zinc

assay

99.95%

assay

99.95%

assay

99.95%

assay

99.95%

form

foil

form

foil

form

foil

form

foil

manufacturer/tradename

Goodfellow 772-508-65

manufacturer/tradename

Goodfellow 705-621-12

manufacturer/tradename

Goodfellow 139-826-43

manufacturer/tradename

Goodfellow 379-172-32

resistivity

5.8 μΩ-cm, 20°C

resistivity

5.8 μΩ-cm, 20°C

resistivity

5.8 μΩ-cm, 20°C

resistivity

5.8 μΩ-cm, 20°C

bp

907 °C (lit.)

bp

907 °C (lit.)

bp

907 °C (lit.)

bp

907 °C (lit.)

mp

420 °C (lit.)

mp

420 °C (lit.)

mp

420 °C (lit.)

mp

420 °C (lit.)

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

(enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service