Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

GF89800599

Tungsten

powder, 250 max. part. size (micron), weight 100 g, min. particle size 50 micron

Synonym(s):

W 006012, W

Sign In to View Organizational & Contract Pricing.

Select a Size

1 EA

$561.00

$561.00


Check Cart for Availability

Request a Bulk Order

About This Item

Empirical Formula (Hill Notation):
W
CAS Number:
Molecular Weight:
183.84
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141747
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI

1S/W

SMILES string

[W]

InChI key

WFKWXMTUELFFGS-UHFFFAOYSA-N

form

powder

manufacturer/tradename

Goodfellow 898-005-99

resistivity

4.9 μΩ-cm, 20°C

weight

100 g

max. part. size

250 (micron)

bp

5660 °C (lit.)

mp

3410 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF94499937GF59398048GF12491793
manufacturer/tradename

Goodfellow 898-005-99

manufacturer/tradename

Goodfellow 944-999-37

manufacturer/tradename

Goodfellow 593-980-48

manufacturer/tradename

Goodfellow 124-917-93

form

powder

form

powder

form

powder

form

powder

mp

3410 °C (lit.)

mp

3410 °C (lit.)

mp

3410 °C (lit.)

mp

3410 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

bp

5660 °C (lit.)

bp

5660 °C (lit.)

bp

5660 °C (lit.)

bp

5660 °C (lit.)

resistivity

4.9 μΩ-cm, 20°C

resistivity

4.9 μΩ-cm, 20°C

resistivity

4.9 μΩ-cm, 20°C

resistivity

4.9 μΩ-cm, 20°C

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Self-heat. 2

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service