Skip to Content
MilliporeSigma

GF91160366

Nickel-chromium alloy, Ni80Cr20

10m wire, 0.25mm diameter, hard

Synonym(s):

NI055130

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$262.00

$262.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
$262.00

About This Item

UNSPSC Code:
11101711
NACRES:
NA.23

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

$262.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

form

wire

manufacturer/tradename

Goodfellow 911-603-66

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF91167059GF59160584GF25167270
form

wire

form

wire

form

wire

form

wire

manufacturer/tradename

Goodfellow 911-603-66

manufacturer/tradename

Goodfellow 911-670-59

manufacturer/tradename

Goodfellow 591-605-84

manufacturer/tradename

Goodfellow 251-672-70

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service