MilliporeSigma
All Photos(2)

GF96917293

Chromium

foil, light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 99.99+%

Synonym(s):
Chromium, CR000110
Empirical Formula (Hill Notation):
Cr
CAS Number:
Molecular Weight:
52.00
MDL number:
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.99%

form

foil

manufacturer/tradename

Goodfellow 969-172-93

resistivity

12.7 μΩ-cm, 20°C

bp

2672 °C (lit.)

mp

1857 °C (lit.)

density

7.14 g/mL at 25 °C (lit.)

SMILES string

[Cr]

InChI

1S/Cr

InChI key

VYZAMTAEIAYCRO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Compare Similar Items

View Full Comparison

1 of 4

This Item
GF24100692GF35390651GF25176472
Chromium foil, light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 99.99+%

GF96917293

Chromium

Chromium foil, light tested, 25x25mm, thickness 0.007mm, permanent polyester support, 99.99+%

GF24100692

Chromium

Chromium foil, light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.99+%

GF35390651

Chromium

Chromium foil, light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.99+%

GF25176472

Chromium

assay

≥99.99%

assay

99.99%

assay

99.99%

assay

99.99%

form

foil

form

foil

form

foil

form

foil

manufacturer/tradename

Goodfellow 969-172-93

manufacturer/tradename

Goodfellow 241-006-92

manufacturer/tradename

Goodfellow 353-906-51

manufacturer/tradename

Goodfellow 251-764-72

resistivity

12.7 μΩ-cm, 20°C

resistivity

12.7 μΩ-cm, 20°C

resistivity

12.7 μΩ-cm, 20°C

resistivity

12.7 μΩ-cm, 20°C

bp

2672 °C (lit.)

bp

2672 °C (lit.)

bp

2672 °C (lit.)

bp

2672 °C (lit.)

mp

1857 °C (lit.)

mp

1857 °C (lit.)

mp

1857 °C (lit.)

mp

1857 °C (lit.)

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

(enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service