Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H206

Sigma-Aldrich

Hecogenin acetate

technical grade, 90%

Synonym(s):

(−)-Hecogenin acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H44O5
CAS Number:
Molecular Weight:
472.66
EC Number:
MDL number:
UNSPSC Code:
12352103

grade

technical grade

assay

90%

optical activity

[α]20/D −6.0°, c = 1 in chloroform

mp

246-248 °C (dec.) (lit.)

SMILES string

[H][C@@]12CC[C@]3([H])[C@]([H])(CC(=O)[C@@]4(C)[C@@]3([H])C[C@]5([H])O[C@]6(CC[C@@H](C)CO6)[C@@H](C)[C@]45[H])[C@@]1(C)CC[C@@H](C2)OC(C)=O

InChI

1S/C29H44O5/c1-16-8-11-29(32-15-16)17(2)26-24(34-29)13-23-21-7-6-19-12-20(33-18(3)30)9-10-27(19,4)22(21)14-25(31)28(23,26)5/h16-17,19-24,26H,6-15H2,1-5H3/t16-,17+,19+,20+,21-,22+,23+,24+,26+,27+,28-,29-/m1/s1

InChI key

CVKZWRTYHCDWTE-RSEFXUKDSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service