All Photos(1)

N16606

Sigma-Aldrich

Nitrocyclohexane

97%

Empirical Formula (Hill Notation):
C6H11NO2
CAS Number:
Molecular Weight:
129.16
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.46 (vs air)

Quality Level

vapor pressure

768 mmHg ( 205 °C)

assay

97%

form

liquid

refractive index

n20/D 1.462 (lit.)

bp

205.5-206 °C/768 mmHg (lit.)

mp

−34 °C (lit.)

density

1.061 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)C1CCCCC1

InChI

1S/C6H11NO2/c8-7(9)6-4-2-1-3-5-6/h6H,1-5H2

InChI key

NJNQUTDUIPVROZ-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service