NCXSPSH850

Sigma-Aldrich

Silver Nanoplates

Silica-Shelled, λmax, 850 nm, NanoXact, 1 mg/mL in sodium bicarbonate (Aqueous 10 mM), Silica (Silanol)

NACRES:
NA.23

form

nanoplates

concentration

1 mg/mL in sodium bicarbonate (Aqueous 10 mM)

λmax

850 nm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Owing to their plasmonic properties nanoparticles of gold and silver find broad range of applications extending from diagnostics to molecular detection, photothermal and microwave-based therapies, and nanotoxicology. Colloidal silver nanoparticles also find applications in antimicrobial coatings, electronics and photovoltaics, and surface enhanced Raman spectroscopy (SERS) whereas the colloidal gold nanoparticles find applications in lateral flow assays (immunochromatographic assays) and as TEM labels and size standards.

Our silver nanoplates (a.k.a. silver nanodisks) exhibit surface plasmon resonance (SPR) effect and, depending upon their diameter and thickness, possess resonance peak between 550nm t0 1050nm. The extremely high absorbing and scattering cross section of these nanoplates make them useful for surface-enhanced Raman scattering (SERS), photovoltaics, molecular detection, and photothermal cancer therapies. The presence of silica shell on silver nanoplates provides robust surface and enhances their stability in variety of solvents. The silica shell offers a versatile surface for attaching functional groups or creating hydrophobic & hydrophilic surfaces. The presence of silica shell on the surface of silver nanoplates also allows their easier deposition onto substrates and incorporation into different solvents, resins, and curable materials while protecting them from aggregation and preserving their optical properties.

Storage and Stability

DO NOT FREEZE. Store at 2-8C and away from light when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.

Legal Information

Product of nanoComposix, Inc.

Pictograms

Environment

Hazard Statements

Precautionary Statements

RIDADR

UN 3082 9 / PGIII

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.