OCNIAC05

Sigma-Aldrich

Iron oxide (III) nanoparticles

5 nm, PEG-Carboxylic acid functionalized, 5 mg/mL in H2O

concentration

5 mg/mL in H2O

particle size

5 nm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Iron oxide nanoparticles are magnetic single crystals and can be used as magnetic resonance imaging contrast reagents for in-vivo imaing , drug delivery vehicle, hyperthermia generator for therapy, adjuvant for vaccine and magnetic sensing probe for in-vitro diagnostics. The PEG-Carboxylic acid functionalized iron oxide nanoparticles are coated with a monolayer of PEG polymer. They offer compact size and low non-specific binding. The reaction group is carboxyl acid which can react with amine containing molecules such as an antibody, peptide, DNA or other small molecules. The available iron oxide core sizes are 5, 10, 20 and 30 nm as namely OCNIAC05, OCNIAC10, OCNIAC20 and OCNIAC30, respectively.

Legal Information

Product of Ocean Nanotech, LLC.

WGK Germany

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.