Key Documents
PL800GA-8
Plas-Labs accordion sleeve with neoprene glove
1 pair, size 8 (for 8 in. glove box ports)
Select a Size
1 PAIR
$895.00
$895.00
Check Cart for Availability
Select a Size
Change View
1 PAIR
$895.00
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
$895.00
Check Cart for Availability
material
neoprene
size
8 (for 8 in. glove box ports)
feature
reusable
packaging
pack of 1 pair
manufacturer/tradename
Plas-Labs 800GA-8
Compare Similar Items
View Full Comparison
Show Differences
1 of 4
This Item
PL800GA-NEO-7PL800GA-NEO-8PL800GA-NEO-9
material
neoprene
material
neoprene
material
neoprene
material
neoprene
manufacturer/tradename
Plas-Labs 800GA-8
manufacturer/tradename
Plas-Labs 800GA-NEO-7
manufacturer/tradename
Plas-Labs 800GA-NEO-8
manufacturer/tradename
Plas-Labs 800GA-NEO-9
feature
reusable
feature
reusable, for use with glove box sleeves
feature
reusable, for use with glove box sleeves
feature
reusable, for use with glove box sleeves
packaging
pack of 1 pair
packaging
pack of 1 pair
packaging
pack of 1 pair
packaging
pack of 1 pair
size
8 (for 8 in. glove box ports)
size
7 (for 8 in. ports)
size
8 (for 8 in. ports)
size
9 (for 8 in. ports)
General description
Plas-Labs glovebox gloves are available as either one-piece or two-piece assemblies with interchangeable sleeves and gloves. This enables personnel to use the same glove box by changing the size or type of glove being used. Choose from four rubber types. Gloves are 15 mil thickness unless noted otherwise.
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
  • White Hypalon® rubber for resistance to abrasion, oxidizing chemicals, ozone and UV light
  • Black neoprene rubber for chemical resistance
  • Black butyl rubber for highest impermeability to water vapor, gases, and toxic fluids
  • Green nitrile rubber for resistance to oil, fuel, and acids
  • Latex rubber for flexibility
Legal Information
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)