Skip to Content
MilliporeSigma

QACIDSET

QuicTPD Acid Optimization Set

greener alternative

Greener Alternative

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.21
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sustainability

Greener Alternative Product

Quality Segment

greener alternative category

storage temp.

2-8°C

General description

This set includes 4 representative partial PROTACs, each provided at 5 micromole quantity.
We are committed to bringing you Greener Alternative Products. This product is a Design for Sustainability (DfS) developed product, which belongs to one of the four categories of greener alternatives. Click here to view its DfS scorecard.

Application

The QuicTPD Acid Optimization set is designed to facilitate high throughput synthesis optimization for researchers working with PROTACs.

Features and Benefits

Two of these PROTACs are tailored to target the CRBN E3 ligase, while the other two are directed towards VHL, incorporating both hydrophobic and hydrophilic linkers. By utilizing this optimization set, customers can explore a variety of reaction conditions—up to 24 or more—before proceeding to the complete screening set. This exploration includes testing different coupling reagents, solvents, and bases, allowing for a comprehensive assessment of the synthesis parameters that influence the efficiency and yield of the desired products.

Other Notes

QuicTPD Acid Screening Set

Legal Information

QuicTPD is a trademark of Merck KGaA, Darmstadt, Germany


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • Pomalidomide-PEG2-CO2H, 95%
    SDS

  • Pomalidomide-C6-CO2H, ≥98%
    SDS

  • VH 032 amide-PEG2-acid, ≥95%
    SDS

  • VH 032 amide-alkyl C5-acid, ≥95%
    SDS

Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library