QBD11436

Sigma-Aldrich

Hexa(-amido-dPEG ®11-MAL)dipentaerythritol

Empirical Formula (Hill Notation):
C214H376N18O97
Molecular Weight:
4753.34

assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: cross-linking reagent
reactivity: thiol reactive

polymer architecture

shape: branched
functionality: homofunctional

shipped in

ambient

storage temp.

−20°C

Inchi Code

1S/C214H376N18O97/c233-189(13-37-227-201(245)1-2-202(227)246)215-25-49-257-61-73-269-85-97-281-109-121-293-133-145-305-157-169-317-175-163-311-151-139-299-127-115-287-103-91-275-79-67-263-55-31-221-195(239)19-43-323-181-213(182-324-44-20-196(240)222-32-56-264-68-80-276-92-104-288-116-128-300-140-152-312-164-176-318-170-158-306-146-134-294-122-110-282-98-86-270-74-62-258-50-26-216-190(234)14-38-228-203(247)3-4-204(228)248,183-325-45-21-197(241)223-33-57-265-69-81-277-93-105-289-117-129-301-141-153-313-165-177-319-171-159-307-147-135-295-123-111-283-99-87-271-75-63-259-51-27-217-191(235)15-39-229-205(249)5-6-206(229)250)187-329-188-214(184-326-46-22-198(242)224-34-58-266-70-82-278-94-106-290-118-130-302-142-154-314-166-178-320-172-160-308-148-136-296-124-112-284-100-88-272-76-64-260-52-28-218-192(236)16-40-230-207(251)7-8-208(230)252,185-327-47-23-199(243)225-35-59-267-71-83-279-95-107-291-119-131-303-143-155-315-167-179-321-173-161-309-149-137-297-125-113-285-101-89-273-77-65-261-53-29-219-193(237)17-41-231-209(253)9-10-210(231)254)186-328-48-24-200(244)226-36-60-268-72-84-280-96-108-292-120-132-304-144-156-316-168-180-322-174-162-310-150-138-298-126-114-286-102-90-274-78-66-262-54-30-220-194(238)18-42-232-211(255)11-12-212(232)256/h1-12H,13-188H2,(H,215,233)(H,216,234)(H,217,235)(H,218,236)(H,219,237)(H,220,238)(H,221,239)(H,222,240)(H,223,241)(H,224,242)(H,225,243)(H,226,244)

InChI key

NSSWOXDMSXMROQ-UHFFFAOYSA-N

Features and Benefits

Hexa(-amido-dPEG ®₁₁-MAL)dipentaerythritol has six connected dPEG spacers that are each capped with a maleimide group which can react with a free thiol.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.